8-(3-Chloro-4-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

124874-500MG 124874-1G 124874-5G 124874-25G
77323-776EA 161.71 USD
77323-776 77346-062 77359-950 77383-692
8-(3-Chloro-4-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(3-Chloro-4-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClFO₃
MW: 286.7264 g/mol
Storage Temperature: Ambient
CAS Number: 2012815-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR