8-(4-Fluoro-2-methylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

124872-500MG 124872-1G 124872-5G 124872-25G
77323-774EA 161.71 USD
77323-774 77346-060 77359-948 77383-690
8-(4-Fluoro-2-methylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(4-Fluoro-2-methylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₅H₁₉FO₃
MW: 266.3079 g/mol
Storage Temperature: Ambient
CAS Number: 1477731-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR