8-(3,4-Difluorophenyl)-1,4-Dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

124868-500MG 124868-1G 124868-5G 124868-25G
77323-772EA 161.71 USD
77323-772 77346-058 77359-946 77383-688
8-(3,4-Difluorophenyl)-1,4-Dioxaspiro[4.5]decan-8-ol ≥95%
8-(3,4-Difluorophenyl)-1,4-Dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₆F₂O₃
MW: 270.2718 g/mol
Storage Temperature: Ambient
CAS Number: 135862-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR