8-(2-Fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

124860-500MG 124860-1G 124860-5G 124860-25G
77323-770EA 161.71 USD
77323-770 77346-056 77359-944 77383-686
8-(2-Fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2-Fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₇FO₃
MW: 252.2813 g/mol
Storage Temperature: Ambient
CAS Number: 218776-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR