1-(3,5-bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
124496-500MG
124496-1G
124496-5G
124496-25G
77323-766EA
130.81
USD
77323-766
77344-320
77359-934
77371-912
1-(3,5-bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanol ≥95%
1-(3,5-bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanol
Precaución: for Research Use Only
Formula:
C₁₀H₅F₉O MW: 312.1317 g/mol Storage Temperature: Ambient |
CAS Number:
1416980-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...