3-Chloro-4-(4-(trifluoromethyl)phenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

124350-500MG 124350-1G 124350-5G
77323-764EA 346.08 USD
77323-764 77347-818 77371-910
3-Chloro-4-(4-(trifluoromethyl)phenyl)pyridine ≥97%
3-Chloro-4-(4-(trifluoromethyl)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₇ClF₃N
MW: 257.639 g/mol
Storage Temperature: Ambient
CAS Number: 1261757-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR