2-hydroxy-5-iodo-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

124233-500MG 124233-1G 124233-5G 124233-25G
77323-760EA 221.45 USD
77323-760 77347-814 77359-932 77383-684
2-hydroxy-5-iodo-3-methylbenzaldehyde ≥97%
2-hydroxy-5-iodo-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.044 g/mol
CAS Number: 83816-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR