1-(5-Fluoropyridin-3-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

124187-500MG 124187-1G 124187-5G 124187-25G
77323-758EA 202.91 USD
77323-758 77347-812 77359-930 77383-682
1-(5-Fluoropyridin-3-yl)cyclopentanol ≥97%
1-(5-Fluoropyridin-3-yl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO
MW: 181.207 g/mol
Storage Temperature: Ambient
CAS Number: 1250764-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR