2-(2-Chloro-6-fluorophenyl)-1,1,1-trifluoropropan-2-ol ≥95%

Supplier: AOBChem USA

Warning

124099-500MG 124099-1G 124099-5G 124099-25G
77323-756EA 223.51 USD
77323-756 77347-810 77359-928 77383-680
2-(2-Chloro-6-fluorophenyl)-1,1,1-trifluoropropan-2-ol ≥95%
2-(2-Chloro-6-fluorophenyl)-1,1,1-trifluoropropan-2-ol

Precaución: for Research Use Only

Formula: C₉H₇ClF₄O
MW: 242.5978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26756224
CAS Number: 1556557-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR