3-Iodo-4-methyl-5-nitrobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

124057-500MG 124057-1G 124057-5G 124057-25G
77323-752EA 182.31 USD
77323-752 77347-094 77359-924 77383-676
3-Iodo-4-methyl-5-nitrobenzoic acid ≥97%
3-Iodo-4-methyl-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆INO₄
Storage Temperature: Ambient
CAS Number: 21312-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR