3-Chloro-4-methyl-5-nitrobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

124052-500MG 124052-1G 124052-5G 124052-25G
77323-750EA 241.02 USD
77323-750 77347-806 77359-922 77383-674
3-Chloro-4-methyl-5-nitrobenzoic acid ≥97%
3-Chloro-4-methyl-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
Storage Temperature: Ambient
CAS Number: 181871-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR