(2,3-difluoro-6-(methylthio)phenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

123422-1G 123422-5G 123422-25G
77341-452EA 86.52 USD
77341-452 77359-914 77371-902
(2,3-difluoro-6-(methylthio)phenyl)trimethylsilane ≥97%
(2,3-difluoro-6-(methylthio)phenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄F₂SSi
Storage Temperature: Ambient
CAS Number: 2918960-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR