2,6-Dibromo-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

123163-500MG 123163-1G 123163-5G 123163-25G
77323-742EA 170.98 USD
77323-742 77346-408 77359-906 77383-666
2,6-Dibromo-N-methoxy-N-methylbenzamide ≥97%
2,6-Dibromo-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉Br₂NO₂
Storage Temperature: Ambient
CAS Number: 2921839-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR