2-Bromo-4-ethyl-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

122792-500MG 122792-1G 122792-5G
77337-930EA 451.14 USD
77337-930 77347-798 77371-892
2-Bromo-4-ethyl-1-methylbenzene ≥97%
2-Bromo-4-ethyl-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.088 g/mol
Storage Temperature: Ambient
CAS Number: 17070-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR