2-Ethyl-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

122787-500MG 122787-1G
77340-552EA 1789.11 USD
77340-552 77347-796
2-Ethyl-5-methylbenzaldehyde ≥97%
2-Ethyl-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.202 g/mol
Storage Temperature: Ambient
CAS Number: 1289169-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR