2-Ethyl-5-methylphenol ≥95%

Supplier: AOBChem USA

Warning

122783-500MG 122783-1G 122783-5G 122783-25G
77323-740EA 202.91 USD
77323-740 77347-794 77359-902 77383-664
2-Ethyl-5-methylphenol ≥95%
2-Ethyl-5-methylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD08691687
CAS Number: 1687-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR