2-Bromo-1-ethyl-4-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
122780-500MG
122780-1G
122780-5G
77339-536EA
594.31
USD
77339-536
77347-792
77371-890
2-Bromo-1-ethyl-4-methylbenzene ≥97%
2-Bromo-1-ethyl-4-methylbenzene
Precaución: for Research Use Only
Formula:
C₉H₁₁Br MW: 199.088 g/mol Storage Temperature: Ambient |
CAS Number:
4748-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...