2-Bromo-1-ethyl-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

122769-500MG 122769-1G 122769-5G 122769-25G
77323-738EA 261.62 USD
77323-738 77347-790 77359-900 77383-662
2-Bromo-1-ethyl-4-methoxybenzene ≥97%
2-Bromo-1-ethyl-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
CAS Number: 852946-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR