3-Ethyl-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

122764-500MG 122764-1G 122764-5G
77339-312EA 576.8 USD
77339-312 77347-788 77371-888
3-Ethyl-4-methylbenzaldehyde ≥97%
3-Ethyl-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
Storage Temperature: Ambient
CAS Number: 461425-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR