4-Bromo-2-ethyl-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

122758-500MG 122758-1G 122758-5G
77337-828EA 448.05 USD
77337-828 77347-786 77371-886
4-Bromo-2-ethyl-1-methylbenzene ≥97%
4-Bromo-2-ethyl-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.088 g/mol
Storage Temperature: Ambient
CAS Number: 1161497-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR