(3-Bromophenyl)(4-Bromophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

122240-1G 122240-5G 122240-25G
77341-494EA 89.61 USD
77341-494 77359-894 77371-878
(3-Bromophenyl)(4-Bromophenyl)methanone ≥97%
(3-Bromophenyl)(4-Bromophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₈Br₂O
Storage Temperature: Ambient
CAS Number: 83699-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR