3-Bromo-4'-chlorobenzophenone ≥97%

Supplier: AOBChem USA

Warning

122239-500MG 122239-1G 122239-5G 122239-25G
77323-736EA 184.37 USD
77323-736 77347-180 77359-892 77383-660
3-Bromo-4'-chlorobenzophenone ≥97%
3-Bromo-4'-chlorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD06201572
CAS Number: 75762-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR