2-Fluoro-6-(p-tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

122144-500MG 122144-1G 122144-25G 122144-5G
77323-730EA 290.46 USD
77323-730 77347-776 77383-654 77371-872
2-Fluoro-6-(p-tolyl)pyridine ≥97%
2-Fluoro-6-(p-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
Storage Temperature: Ambient
CAS Number: 1245646-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR