2,6-Dibromo-3,4-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

121983-500MG 121983-1G 121983-5G 121983-25G
77323-728EA 267.8 USD
77323-728 77347-774 77359-888 77383-652
2,6-Dibromo-3,4-dimethylphenol ≥97%
2,6-Dibromo-3,4-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
Storage Temperature: Ambient
CAS Number: 22802-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR