(2,4,6-trifluoro-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

121930-500MG 121930-1G 121930-5G 121930-25G
77323-726EA 117.42 USD
77323-726 77343-538 77359-884 77371-866
(2,4,6-trifluoro-3-methoxyphenyl)methanol ≥97%
(2,4,6-trifluoro-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
MW: 192.135 g/mol
Storage Temperature: Ambient
CAS Number: 1804443-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR