2-(3-Bromo-2-chloro-4-fluorophenyl)-1,3-dioxolane

Supplier: AOBChem USA

Warning

121852-500MG 121852-1G 121852-5G 121852-25G
77323-718EA 241.02 USD
77323-718 77347-772 77359-874 77383-644
2-(3-Bromo-2-chloro-4-fluorophenyl)-1,3-dioxolane
2-(3-Bromo-2-chloro-4-fluorophenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₉H₇BrClFO₂
MW: 281.506 g/mol
Storage Temperature: Ambient
CAS Number: 2918952-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR