3-Bromo-4-fluoro-5-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

121851-500MG 121851-1G 121851-5G
77323-716EA 328.57 USD
77323-716 77347-770 77371-862
3-Bromo-4-fluoro-5-methylbenzamide ≥97%
3-Bromo-4-fluoro-5-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
Storage Temperature: Ambient
CAS Number: 2091247-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR