(2-methylnaphthalen-1-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

12160-1G 12160-5G 12160-25G
77341-474EA 89.61 USD
77341-474 77359-870 77371-858
(2-methylnaphthalen-1-yl)boronic acid ≥97%
(2-methylnaphthalen-1-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₂
Storage Temperature: Ambient
MDL Number: MFCD03452758
CAS Number: 103989-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR