1-Bromo-2,3,5-trifluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

121585-500MG 121585-1G 121585-5G
77336-228EA 383.16 USD
77336-228 77347-768 77371-856
1-Bromo-2,3,5-trifluoro-4-iodobenzene ≥97%
1-Bromo-2,3,5-trifluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆HBrF₃I
MW: 336.876 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32669845
CAS Number: 2514941-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR