4-Bromo-2,3,6-trifluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

121582-500MG 121582-1G 121582-5G 121582-25G
77323-714EA 146.26 USD
77323-714 77345-188 77359-868 77383-642
4-Bromo-2,3,6-trifluorobenzoic acid ≥97%
4-Bromo-2,3,6-trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06203571
CAS Number: 415965-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR