1-Bromo-3-chloro-5-ethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

121254-500MG 121254-1G 121254-5G 121254-25G
77323-712EA 181.28 USD
77323-712 77346-880 77359-866 77383-640
1-Bromo-3-chloro-5-ethoxybenzene ≥95%
1-Bromo-3-chloro-5-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
CAS Number: 1881329-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR