4-Bromo-2-fluoro-5-methoxyaniline ≥97%

Supplier: AOBChem USA

Warning

120952-500MG 120952-1G 120952-5G
77323-710EA 317.24 USD
77323-710 77347-764 77371-852
4-Bromo-2-fluoro-5-methoxyaniline ≥97%
4-Bromo-2-fluoro-5-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD22199648
CAS Number: 108310-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR