2,6-Difluoro-3-(trimethylsilyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

120939-1G 120939-5G 120939-25G
77342-090EA 112.27 USD
77342-090 77359-862 77371-848
2,6-Difluoro-3-(trimethylsilyl)benzaldehyde ≥97%
2,6-Difluoro-3-(trimethylsilyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂OSi
MW: 214.284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784678
CAS Number: 2901103-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR