2-Bromo-3,6-difluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1,4-difluoro-3-iodobenzene

120928-500MG 120928-1G 120928-5G 120928-25G
77323-708EA 174.07 USD
77323-708 77346-476 77359-860 77383-638
2-Bromo-3,6-difluoroiodobenzene ≥97%
2-Bromo-3,6-difluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂I
MW: 318.885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922621
CAS Number: 1208074-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR