2-hydroxy-3-(trimethylsilyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

120907-500MG 120907-1G 120907-5G 120907-25G
77323-706EA 132.87 USD
77323-706 77344-410 77359-858 77383-636
2-hydroxy-3-(trimethylsilyl)benzoic acid ≥97%
2-hydroxy-3-(trimethylsilyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃Si
Storage Temperature: Ambient
MDL Number: MFCD34784674
CAS Number: 2901110-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR