2-Bromo-3-chloro-6-fluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-chloro-4-fluoro-3-iodobenzene

120893-500MG 120893-1G 120893-25G 120893-5G
77323-704EA 285.31 USD
77323-704 77347-762 77383-634 77371-846
2-Bromo-3-chloro-6-fluoroiodobenzene ≥97%
2-Bromo-3-chloro-6-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
MW: 335.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29037266
CAS Number: 1935178-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR