3-Ethoxy-4-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

120841-500MG 120841-1G 120841-5G 120841-25G
77323-702EA 158.62 USD
77323-702 77345-750 77359-856 77383-632
3-Ethoxy-4-methoxybenzamide ≥97%
3-Ethoxy-4-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.215 g/mol
Storage Temperature: Ambient
CAS Number: 247569-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR