5-Methoxy-8-methylquinoline ≥97%

Supplier: AOBChem USA

Warning

120745-500MG 120745-1G 120745-25G 120745-5G
77323-700EA 301.79 USD
77323-700 77347-760 77383-630 77371-844
5-Methoxy-8-methylquinoline ≥97%
5-Methoxy-8-methylquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
Storage Temperature: Ambient
CAS Number: 868668-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR