2,5-Dibromo-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

120658-500MG 120658-1G 120658-5G 120658-25G
77323-698EA 120.51 USD
77323-698 77343-860 77359-850 77371-838
2,5-Dibromo-3-(trifluoromethyl)benzaldehyde ≥97%
2,5-Dibromo-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃Br₂F₃O
MW: 331.912 g/mol
Storage Temperature: Ambient
CAS Number: 2166945-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR