2-Bromo-5-nitro-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

120657-500MG 120657-1G 120657-5G 120657-25G
77323-696EA 112.27 USD
77323-696 77343-320 77359-848 77371-836
2-Bromo-5-nitro-3-(trifluoromethyl)benzaldehyde ≥97%
2-Bromo-5-nitro-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃BrF₃NO₃
Storage Temperature: Ambient
CAS Number: 2383482-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR