N-(2-Bromo-4-iodophenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

120641-500MG 120641-1G 120641-5G 120641-25G
77323-694EA 150.38 USD
77323-694 77345-376 77359-846 77383-628
N-(2-Bromo-4-iodophenyl)acetamide ≥97%
N-(2-Bromo-4-iodophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇BrINO
MW: 339.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13186695
CAS Number: 474966-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR