(Z)-1-(4-bromophenyl)propan-1-one oxime ≥97%

Supplier: AOBChem USA

Warning

120620-500MG 120620-1G 120620-5G
77336-852EA 394.49 USD
77336-852 77347-758 77371-832
(Z)-1-(4-bromophenyl)propan-1-one oxime ≥97%
(Z)-1-(4-bromophenyl)propan-1-one oxime

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MW: 228.086 g/mol
Storage Temperature: Ambient
CAS Number: 153457-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR