(E)-1-(2-bromophenyl)ethanone oxime ≥97%

Supplier: AOBChem USA

Warning

120618-1G 120618-5G 120618-25G
77342-100EA 113.3 USD
77342-100 77359-842 77371-830
(E)-1-(2-bromophenyl)ethanone oxime ≥97%
(E)-1-(2-bromophenyl)ethanone oxime

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MW: 214.059 g/mol
Storage Temperature: Ambient
CAS Number: 1040191-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR