4-(Cyclopent-1-en-1-yl)-1-ethoxy-2-fluorobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
120345-500MG
120345-1G
120345-5G
120345-25G
77323-688EA
127.72
USD
77323-688
77344-230
77359-832
77371-822
4-(Cyclopent-1-en-1-yl)-1-ethoxy-2-fluorobenzene ≥97%
4-(Cyclopent-1-en-1-yl)-1-ethoxy-2-fluorobenzene
Precaución: for Research Use Only
Formula:
C₁₃H₁₅FO MW: 206.256 g/mol Storage Temperature: Ambient |
CAS Number:
2918967-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...