(3-Bromo-2-(methylthio)phenyl)(ethyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

120298-500MG 120298-1G 120298-5G 120298-25G
77323-686EA 119.48 USD
77323-686 77343-658 77359-830 77371-820
(3-Bromo-2-(methylthio)phenyl)(ethyl)sulfane ≥97%
(3-Bromo-2-(methylthio)phenyl)(ethyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁BrS₂
Storage Temperature: Ambient
MDL Number: MFCD34784656
CAS Number: 2918845-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR