2,6-Dichloro-3,5-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-dichloro-3,5-dimethoxybenzoic acid

120285-500MG 120285-1G 120285-5G 120285-25G
77323-684EA 120.51 USD
77323-684 77343-858 77359-828 77371-818
2,6-Dichloro-3,5-dimethoxybenzoic acid ≥97%
2,6-Dichloro-3,5-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₄
MW: 251.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18430088
CAS Number: 75177-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR