(4-Bromo-1,2-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

120183-500MG 120183-1G 120183-5G 120183-25G
77323-680EA 86.52 USD
77323-680 77342-264 77359-824 77371-814
(4-Bromo-1,2-phenylene)bis(methylsulfane) ≥97%
(4-Bromo-1,2-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₉BrS₂
Storage Temperature: Ambient
CAS Number: 138709-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR