2-Methyl-4-(methylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

120069-500MG 120069-1G 120069-5G 120069-25G
77323-678EA 211.15 USD
77323-678 77347-750 77359-822 77383-622
2-Methyl-4-(methylthio)benzonitrile ≥97%
2-Methyl-4-(methylthio)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NS
Storage Temperature: Ambient
CAS Number: 1190948-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR