2,6-Dibromobenzene-1,4-diol ≥95%

Supplier: AOBChem USA

Warning

120011-500MG 120011-1G 120011-5G 120011-25G
77323-676EA 131.84 USD
77323-676 77344-382 77359-820 77383-620
2,6-Dibromobenzene-1,4-diol ≥95%
2,6-Dibromobenzene-1,4-diol

Precaución: for Research Use Only

Formula: C₆H₄Br₂O₂
Storage Temperature: Ambient
CAS Number: 3333-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR