Ethyl 3-bromo-2-hydroxybenzoate ≥95%

Supplier: AOBChem USA

Warning

120008-500MG 120008-1G 120008-5G 120008-25G
77323-674EA 113.3 USD
77323-674 77343-352 77359-818 77371-812
Ethyl 3-bromo-2-hydroxybenzoate ≥95%
Ethyl 3-bromo-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566799
CAS Number: 314240-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR